Stampa…

Anti-Acetyl Coenzyme A carboxylase alpha Rabbit Monoclonal Antibody [clone: EPR23235-147]

Fornitore: Abcam
AB272704-100 AB272704-1
ABCAAB272704-100EA 1020 EUR
ABCAAB272704-100 ABCAAB272704-1
Anti-Acetyl Coenzyme A carboxylase alpha Rabbit Monoclonal Antibody [clone: EPR23235-147]
Anticorpi
Rabbit monoclonal [EPR23235-147] to Acetyl Coenzyme A carboxylase alpha - BSA and Azide free.

Type: Primary
Antigen: Acetyl Coenzyme A carboxylase alpha
Clonality: Monoclonal
Clone: EPR23235-147
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR