Stampa…

Anti-Zonula occludens protein 3/ZO3 Rabbit Monoclonal Antibody [clone: EPR13414]

Fornitore: Abcam
AB181991-100 AB181991-1ML
ABCAAB181991-100EA 475 EUR
ABCAAB181991-100 ABCAAB181991-1ML
Anti-Zonula occludens protein 3/ZO3 Rabbit Monoclonal Antibody [clone: EPR13414]
Anticorpi
Rabbit monoclonal [EPR13414] to Zonula occludens protein 3/ZO3 - Plasma Membrane Marker.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Zonula occludens protein 3/ZO3
Clonality: Monoclonal
Clone: EPR13414
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR