Stampa…
Capillary columns, PERMABOND® FFAP
Fornitore: MACHEREY-NAGEL
723116.10 723116.25 723116.30 723116.50 723116.60 723180.10 723180.20 723181.10 723341.25 723341.30 723341.50 723341.60 723344.10 723344.25 723344.30 723344.50 723344.60 723356.25 723356.50 723555.10 723555.25 723555.50 723830.10 723830.25 723830.50 723936.25 723936.50
MANA723116.10EA 273 EUR
MANA723116.10 MANA723116.25 MANA723116.30 MANA723116.50 MANA723116.60 MANA723180.10 MANA723180.20 MANA723181.10 MANA723341.25 MANA723341.30 MANA723341.50 MANA723341.60 MANA723344.10 MANA723344.25 MANA723344.30 MANA723344.50 MANA723344.60 MANA723356.25 MANA723356.50 MANA723555.10 MANA723555.25 MANA723555.50 MANA723830.10 MANA723830.25 MANA723830.50 MANA723936.25 MANA723936.50
Capillary columns, PERMABOND® FFAP
Colonne cromatografiche
Polar GC phase.
  • Recommended for fatty acid methyl esters (FAMEs) and free carboxylic acids
  • Maximum temperature for isothermal operation 220 °C (0,53 mm Int.Ø columns: 200 °C)
  • Maximum temperature for short isotherms in a temperature program 240 °C (0,53 mm Int.Ø columns: 220 °C)
Polyethylene glycol 2-nitroterephthalate phase.
USP code: close equivalent to G25 / G35.