Stampa…
Palladio (II) nitrato (10% Pd) w/w aqueous solution
Fornitore: Thermo Fisher Scientific
Pericolo
12621.NB 12621.ND 12621.NH
12621.NBEA 590 EUR
12621.NB 12621.ND 12621.NH
Palladio (II) nitrato (10% Pd) w/w aqueous solution
Palladio (II) nitrato
CAS No.: 10102-05-3 Molecular Formula: Pd(NO3)2 Formula Weight: 230.43 Physical Form: Liquid Appearance: Brown to yellow to orange UN No.: 3264 MDL No.: MFCD00011169
Formula: N₂O₆Pd
Peso molecolare: 230.43 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011169
CAS Number: 10102-05-3
EINECS: 233-265-8
UN: 3264
ADR: 8 II
Merck Index: 12,07124