2,1-Benzisoxazole ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Benzo[c]isoxazole
A2337-1G
A2337-5G
TCIAA2337-1GEA
49
EUR
TCIAA2337-1G
TCIAA2337-5G
2,1-Benzisoxazole ≥98.0% (by GC)
2,1-Benzisoxazole
Formula:
C₇H₅NO Peso molecolare: 119,1207 g/mol Punto di ebollizione: 101°C Flash Pt: 94°C |
MDL Number:
MFCD00005806 CAS Number: 271-58-9 EINECS: 205-980-5 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...