(±)-Jasmonic acid
Fornitore: Cayman Chemical
Formula:
C₁₂H₁₈O₃ |
MDL Number:
MFCD04221274 CAS Number: 77026-92-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...