Thioisonicotinamide ≥97.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Pyridine-4-carbothioamide
T0764-25G
T0764-5G
TCIAT0764-25GEA
65
EUR
TCIAT0764-25G
TCIAT0764-5G
Thioisonicotinamide ≥97.0% (by titrimetric analysis)
Thioisonicotinamide
Formula:
C₆H₆N₂S Peso molecolare: 138,19 g/mol |
MDL Number:
MFCD00006437 CAS Number: 2196-13-6 EINECS: 218-592-6 |
Risultati del test di specificazione
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Solubility in hot Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...