Sodium isonicotinate ≥99.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Isonicotinic acid sodium salt, Sodium Isonicotinate, Pyridine-4-carboxylic acid sodium salt, Sodium Pyridine-4-carboxylate
P0991-25G
P0991-250G
TCIAP0991-25GEA
36
EUR
TCIAP0991-25G
TCIAP0991-250G
Sodium isonicotinate ≥99.0% (by HPLC, titration analysis)
Sodium isonicotinate
Formula:
C₆H₄NNaO₂ Peso molecolare: 145,0912 g/mol |
MDL Number:
MFCD00064868 CAS Number: 16887-79-9 EINECS: 200-215-1 |
Risultati del test di specificazione
Appearance | White powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Purity (Nonaqueous Titration) | min. 99.0 % |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...