Sodium nicotinate ≥98.0% (by titrimetric analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₆H₄NNaO₂ |
MDL Number:
MFCD00064352 CAS Number: 54-86-4 EINECS: 200-215-1 |
|
Risultati del test di specificazione
Appearance | White powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...