2-Methoxy-5-pyridineboronic acid (contains varying amounts of Anhydride)
Fornitore: TCI
|
Attenzione
|
Synonyms:
(6-Methoxypyridin-3-yl)boronic acid
Formula:
C₆H₈BNO₃ Punto di fusione: 138°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD02093044 CAS Number: 163105-89-3 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 113.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...