Stampa…
4-(4-Fluorophenoxy)benzaldehyde ≥98.0% (by GC)
Fornitore: TCI EUROPE
Attenzione
F0537-5G F0537-1G
TCIAF0537-5GEA 229 EUR
TCIAF0537-5G TCIAF0537-1G
4-(4-Fluorophenoxy)benzaldehyde ≥98.0% (by GC)
4-(4-Fluorophenoxy)benzaldehyde
Formula: C₁₃H₉FO₂
Peso molecolare: 216,21 g/mol
MDL Number: MFCD01631896
CAS Number: 137736-06-2
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66104764270dd998f3045d62e62784b222a5ba3b&vid=66104764270dd998f3045d62e62784b222a5ba3b&prid=web&clid=SAPDAM
Visualizza i codici
Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 75.0 to 79.0 deg-C