Bismuth(III) 2-ethylhexanoate
Fornitore: Thermo Scientific
|
Attenzione
|
Synonyms:
Bismuth 2-ethylhexanoate
36710.14
36710.22
36710.06
36710.14EA
134
EUR
36710.14
36710.22
36710.06
Bismuth(III) 2-ethylhexanoate
Bismuth(III) 2-ethylhexanoate
CAS No.: 67874-71-9
Molecular Formula: C24H45BiO6
Formula Weight: 638.61
Physical Form: Viscous liquid
MDL No.: MFCD00043803
Formula:
C₂₄H₄₅BiO₆ Peso molecolare: 638,598 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00043803 CAS Number: 67874-71-9 EINECS: 267-499-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...