N-Lignoceroyl taurine
Fornitore: Cayman Chemical
10007286-1
10007286-10
10007286-5
CAYM10007286-1EA
172
EUR
CAYM10007286-1
CAYM10007286-10
CAYM10007286-5
N-Lignoceroyl taurine
N-Lignoceroyl taurine
Formula:
C₂₆H₅₃NO₄S |
CAS Number:
807370-75-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...