2,4,4,6-Tetrabromo-2,5-cyclohexadienone ≥97.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2,4,4,6-Tetrabromo-2,5-cyclohexadien-1-one
T1235-25G
T1235-5G
TCIAT1235-25GEA
395
EUR
TCIAT1235-25G
TCIAT1235-5G
2,4,4,6-Tetrabromo-2,5-cyclohexadienone ≥97.0% (by titrimetric analysis)
2,4,4,6-Tetrabromo-2,5-cyclohexadienone
Formula:
C₆H₂Br₄O Peso molecolare: 409,700 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00001589 CAS Number: 20244-61-5 EINECS: 243-638-7 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (Iodometric Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...