Stampa…

3-Bromochromone ≥98.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: 3-Bromo-4H-chromen-4-one

B3334-25G B3334-5G
TCIAB3334-25GEA 384 EUR
TCIAB3334-25G TCIAB3334-5G
3-Bromochromone ≥98.0% (by GC)
3-Bromochromone
Formula: C₉H₅BrO₂
Peso molecolare: 225.05 g/mol
MDL Number: MFCD00017337
CAS Number: 49619-82-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c3ef734653f02ba2b53f6deef72f205c5d5f39c&vid=4c3ef734653f02ba2b53f6deef72f205c5d5f39c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 95.0 to 99.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR