Stampa…

Ethoxyquin ≥90.0% (by GC) antioxidant for rubber

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-Ethoxy-1,2-dihydro-2,2,4-trimethylquinoline, 6-Ethoxy-2,2,4-trimethyl-1,2-dihydroquinoline, 1,2-Dihydro-6-ethoxy-2,2,4-trimethylquinoline, Ethoxyquin

E0237-25G E0237-500G
TCIAE0237-25GEA 16 EUR
TCIAE0237-25G TCIAE0237-500G
Ethoxyquin ≥90.0% (by GC) antioxidant for rubber
Ethoxyquin
Formula: C₁₄H₁₉NO
Punto di ebollizione: 161°C
Flash Pt: 137°C
MDL Number: MFCD00023883
CAS Number: 91-53-2
EINECS: 202-075-7
UN: 3082
ADR: 9,III
Merck Index: 14,03753
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32554e36573c41eee11d0f7519387244212ffc2c&vid=40ff4432325e2676b7284f041686e08721eada08&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Red to Dark blue to Black clear liquid to cloudy liquid
Purity (GC) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR