Stampa…

Betaine, anidro ≥97.0% (by titrimetric analysis)

Fornitore: TCI

B0455-25G B0455-500G B0455-100G
TCIAB0455-25GEA 16 EUR
TCIAB0455-25G TCIAB0455-500G TCIAB0455-100G
Betaine, anidro ≥97.0% (by titrimetric analysis)
Betaine
Formula: C₅H₁₁NO₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00012123
CAS Number: 107-43-7
EINECS: 203-490-6
Merck Index: 14,01179
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b92ecc4897b3fad4ff15e73c2e3dcc3dacd4bdf&vid=14a492a26597a0ed98ecd870e861577691f27e40&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR