N,N,N',N',N''-Pentamethyldiethylenetriamine ≥99.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
N1-(2-(Dimethylamino)ethyl)-N1,N2,N2-trimethylethane-1,2-diamine, 1,1,4,7,7-Pentamethyldiethylenetriamine
P0881-25ML
P0881-500ML
P0881-100ML
TCIAP0881-25MLEA
19
EUR
TCIAP0881-25ML
TCIAP0881-500ML
TCIAP0881-100ML
N,N,N',N',N''-Pentamethyldiethylenetriamine ≥99.0% (by GC)
N,N,N',N',N''-Pentamethyldiethylenetriamine
Formula:
C₉H₂₃N₃ Peso molecolare: 173,30 g/mol Punto di ebollizione: 86°C Punto di fusione: -20°C Flash Pt: 72°C |
MDL Number:
MFCD00014876 CAS Number: 3030-47-5 EINECS: 221-201-1 UN: 2922 ADR: 8,II |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...