Stampa…

2-Nitrophenyl octyl ether ≥98.0% (by GC) matrix for FABMS and liquid SIMS

Fornitore: TCI EUROPE

Synonyms: 1-Nitro-2-(n-octyloxy)benzene, 1-Nitro-2-(octyloxy)benzene, 2-(n-Octyloxy)nitrobenzene

S0380-1G S0380-10G
TCIAS0380-1GEA 17 EUR
TCIAS0380-1G TCIAS0380-10G
2-Nitrophenyl octyl ether ≥98.0% (by GC) matrix for FABMS and liquid SIMS
2-Nitrophenyl octyl ether
Formula: C₁₄H₂₁NO₃
Peso molecolare: 251,32 g/mol
Punto di ebollizione: 198°C
MDL Number: MFCD00014693
CAS Number: 37682-29-4
EINECS: 253-623-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7f8eb7e84565f1e44c976f4cc20dbbebbb0f8d2&vid=57ca3217c561dea3665d6d23033baf74388f0292&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR