Potassium methyl malonate ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Monomethyl monopotassium malonate, Monomethyl malonate potassium salt
M2149-25G
M2149-250G
TCIAM2149-25GEA
26
EUR
TCIAM2149-25G
TCIAM2149-250G
Potassium methyl malonate ≥98.0% (by titrimetric analysis)
Potassium methyl malonate
Formula:
C₄H₅KO₄ Peso molecolare: 156,18 g/mol Punto di fusione: 162°C |
MDL Number:
MFCD00014021 CAS Number: 38330-80-2 EINECS: 253-886-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Ion exchange titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...