Stampa…

2,5-Dimercapto-1,3,4-thiadiazole ≥95.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 1,3,4-Thiadiazole-2,5-dithiol

B0593-25G B0593-500G
TCIAB0593-25GEA 24 EUR
TCIAB0593-25G TCIAB0593-500G
2,5-Dimercapto-1,3,4-thiadiazole ≥95.0% (by titrimetric analysis)
2,5-Dimercapto-1,3,4-thiadiazole
Formula: C₂H₂N₂S₃
Peso molecolare: 150,25 g/mol
MDL Number: MFCD00003103
CAS Number: 1072-71-5
EINECS: 214-014-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6959342ba5af67f37f8bbdd94edfc6bcfd25fa0&vid=d6959342ba5af67f37f8bbdd94edfc6bcfd25fa0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Green powder to crystal
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR