Stampa…

(R)-(-)-Thiazolidine-4-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: H-Thz-OH, L(-)-Thiazolidine-4-carboxylic acid

T0219-25G T0219-500G
TCIAT0219-25GEA 37 EUR
TCIAT0219-25G TCIAT0219-500G
(R)-(-)-Thiazolidine-4-carboxylic acid ≥98.0% (by HPLC, titration analysis)
(R)-(-)-Thiazolidine-4-carboxylic acid
Formula: C₄H₇NO₂S
Peso molecolare: 133,17 g/mol
MDL Number: MFCD00005212
CAS Number: 34592-47-7
EINECS: 252-106-3
UN: 2811
ADR: 6.1,III
Merck Index: 14,09445
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=511f85689a1208048f18f186d095c866ac8a95b3&vid=511f85689a1208048f18f186d095c866ac8a95b3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -197.0 to -202.0 deg (C=1, 1mol/L NaOH sol.)
Solubility in hot Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR