Stampa…

3,5-Dimethyl-4-iodo-1H-pyrazole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3,5-Dimethyl-4-iodopyrazole

I0771-1G I0771-5G
TCIAI0771-1GEA 16 EUR
TCIAI0771-1G TCIAI0771-5G
3,5-Dimethyl-4-iodo-1H-pyrazole ≥98.0% (by GC)
3,5-Dimethyl-4-iodo-1H-pyrazole
Formula: C₅H₇IN₂
Peso molecolare: 222,03 g/mol
MDL Number: MFCD00040247
CAS Number: 2033-45-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8866983f01d3a326a4532180db12a8c5bbcb5c7a&vid=8866983f01d3a326a4532180db12a8c5bbcb5c7a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 138.0 to 142.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR