Stampa…

2,2,6,6-Tetramethylpiperidinooxy (TEMPO) ≥98.0% (by GC, titration analysis) free radical

Fornitore: TCI EUROPE
Pericolo

Synonyms: TEMPO

T1560-25G T1560-5G
TCIAT1560-25GEA 72 EUR
TCIAT1560-25G TCIAT1560-5G
2,2,6,6-Tetramethylpiperidinooxy (TEMPO) ≥98.0% (by GC, titration analysis) free radical
2,2,6,6-Tetramethylpiperidinooxy (TEMPO)
Formula: C₉H₁₈NO
Peso molecolare: 156,25 g/mol
Flash Pt: 67°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00009599
CAS Number: 2564-83-2
EINECS: 219-888-8
UN: 3263
ADR: 8,II
Merck Index: 14,09140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78dd156751cb886b0659ff709f3adf5a44dc6867&vid=bc5669be540fcbbe09883c3c00e69e15bf5b72e2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Orange to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Iodometric Titration) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR