Stampa…

1-Bromo-4-phenylbutane ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

P1388-5G P1388-25G
TCIAP1388-5GEA 93 EUR
TCIAP1388-5G TCIAP1388-25G
1-Bromo-4-phenylbutane ≥97.0% (by GC)
1-Bromo-4-phenylbutane
Formula: C₁₀H₁₃Br
MDL Number: MFCD00154988
CAS Number: 13633-25-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c09565b585b882db7e351a9538315c41ba8955b7&vid=c09565b585b882db7e351a9538315c41ba8955b7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR