Stampa…

(R)-(+)-Hydroxypyrrolidine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (R)-Pyrrolidin-3-ol, (R)-(+)-3-Pyrrolidinol, (R)-(+)-3-Hydroxypyrrolidine

P1608-1G P1608-5G
TCIAP1608-1GEA 29 EUR
TCIAP1608-1G TCIAP1608-5G
(R)-(+)-Hydroxypyrrolidine ≥98.0% (by GC)
(R)-(+)-Hydroxypyrrolidine
Formula: C₄H₉NO
Peso molecolare: 87,12 g/mol
Punto di ebollizione: 70°C
MDL Number: MFCD00145220
CAS Number: 2799-21-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a1552c0f4aea5bebbe1da4dcbcf315cc0a6dcc0&vid=0a1552c0f4aea5bebbe1da4dcbcf315cc0a6dcc0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 99.0 ee%
Purity (Nonaqueous Titration) min. 97.0 %
Specific rotation [a]20/D +4.5 to +7.0 deg (C=3, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR