Stampa…
2-Chloro-3-formylpyridine ≥98.0% (by GC)
Fornitore: TCI EUROPE
Attenzione
Synonyms: 2-Chloropyridine-3-carboxaldehyde, 2-Chloronicotinaldehyde, 2-Chloro-3-pyridinecarboxaldehyde
C2487-1G C2487-5G
TCIAC2487-1GEA 22 EUR
TCIAC2487-1G TCIAC2487-5G
2-Chloro-3-formylpyridine ≥98.0% (by GC)
2-Chloro-3-formylpyridine
Formula: C₆H₄ClNO
Peso molecolare: 141,56 g/mol
MDL Number: MFCD01315308
CAS Number: 36404-88-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7231f17fb002b3d074d36638f7eb06cae24d6ae3&vid=7231f17fb002b3d074d36638f7eb06cae24d6ae3&prid=web&clid=SAPDAM
Visualizza i codici
Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 54.0 deg-C