Stampa…

2-Bromo-5-formylpyridine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-Bromonicotinaldehyde, 2-Bromopyridine-5-carbaldehyde, 2-Bromopyridine-5-carboxaldehyde, 6-Bromo-3-pyridinecarboxaldehyde

B3491-1G B3491-5G
TCIAB3491-1GEA 37 EUR
TCIAB3491-1G TCIAB3491-5G
2-Bromo-5-formylpyridine ≥98.0% (by GC)
2-Bromo-5-formylpyridine
Formula: C₆H₄BrNO
Peso molecolare: 186,01 g/mol
MDL Number: MFCD04115419
CAS Number: 149806-06-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 100.0 to 104.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR