Stampa…

Dichloro(methyl)(phenyl)silane ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Methylphenyldichlorosilane

P0790-25G P0790-250G
TCIAP0790-25GEA 38 EUR
TCIAP0790-25G TCIAP0790-250G
Dichloro(methyl)(phenyl)silane ≥98.0% (by GC)
Dichloro(methyl)(phenyl)silane
Formula: C₇H₈Cl₂Si
Peso molecolare: 191,13 g/mol
Punto di ebollizione: 205°C
Flash Pt: 82°C
MDL Number: MFCD00000490
CAS Number: 149-74-6
EINECS: 205-746-2
UN: 2437
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b289c5b7768fd71344701714a3b7bb1eaf6d552f&vid=b289c5b7768fd71344701714a3b7bb1eaf6d552f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR