Stampa…

α-Benzoinoxime ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Hydroxy-1,2-diphenylethanone oxime, α-Benzoin oxime

B0081-25G B0081-500G
TCIAB0081-25GEA 39 EUR
TCIAB0081-25G TCIAB0081-500G
α-Benzoinoxime ≥98.0% (by HPLC, total nitrogen)
α-Benzoinoxime
Formula: C₁₄H₁₃NO₂
Peso molecolare: 227,26 g/mol
MDL Number: MFCD00004501
CAS Number: 441-38-3
EINECS: 207-127-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69306df090b6b9a3d73b63ff302abe8b0178be52&vid=69306df090b6b9a3d73b63ff302abe8b0178be52&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 150.0 to 155.0 deg-C
Solubility in hot EtOH almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR