4-(Methylthio)phenylboronic acid (contains varying amounts of Anhydride)
Fornitore: TCI
|
Attenzione
|
Synonyms:
4-(Methylthio)benzeneboronic acid, 4-Thioanisoleboronic acid
Formula:
C₇H₉BO₂S Punto di fusione: 213°C |
MDL Number:
MFCD00093410 CAS Number: 98546-51-1 |
|
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (Neutralization titration) | 98.0 to 112.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...