Stampa…

3-Acetoxyindole ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Indolyl Acetate, Indoxyl acetate, 3-Indoxyl acetate, 3-Indolyl acetate

A0068-1G A0068-5G
TCIAA0068-1GEA 71 EUR
TCIAA0068-1G TCIAA0068-5G
3-Acetoxyindole ≥98.0% (by HPLC, total nitrogen)
3-Acetoxyindole
Formula: C₁₀H₉NO₂
Peso molecolare: 175,19 g/mol
MDL Number: MFCD00014561
CAS Number: 608-08-2
EINECS: 210-154-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1093a8f1d8a57262963d72c30e066a65eaab0bbb&vid=1093a8f1d8a57262963d72c30e066a65eaab0bbb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 126.0 to 131.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR