Sanger Reagente ≥99.0% (by GC) for HPLC labeling
Fornitore: TCI
|
Pericolo
|
Synonyms:
1,3-Dinitro-4-fluorobenzene, 2,4-Dinitrofluorobenzene, 1-Fluoro-2,4-dinitrobenzene
Formula:
C₆H₃FN₂O₄ Punto di ebollizione: 296°C Punto di fusione: 26°C Densità: 1,48 g/cm³ (20 °C) Flash Pt: ≥ 100 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00007056 CAS Number: 70-34-8 EINECS: 200-734-3 UN: 2811 ADR: 6.1,II Merck Index: 14,04172 |
|
Risultati del test di specificazione
Appearance | White or Colorless to Yellow to Green powder to lump to clear liquid |
Purity (GC) | min. 99.0 % |
Effect test of HPLC derivatization | Effective as labeling agent for Propylamine, Diethylamine |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...