Stampa…

2-Hydroxy-5-(trifluoromethyl)pyridine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-(Trifluoromethyl)pyridin-2-ol

T2539-25G T2539-5G
TCIAT2539-25GEA 164 EUR
TCIAT2539-25G TCIAT2539-5G
2-Hydroxy-5-(trifluoromethyl)pyridine ≥98.0% (by GC, titration analysis)
2-Hydroxy-5-(trifluoromethyl)pyridine
Formula: C₆H₄F₃NO
Peso molecolare: 163,1 g/mol
MDL Number: MFCD00042315
CAS Number: 33252-63-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db160f994e3b923afa3d6af49fb4092b0144b6aa&vid=db160f994e3b923afa3d6af49fb4092b0144b6aa&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 147.0 to 151.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR