3-Bromo-5-chloro-2-hydroxybenzaldehyde ≥96.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-Bromo-5-chlorosalicylaldehyde
B1948-25G
B1948-5G
TCIAB1948-25GEA
252
EUR
TCIAB1948-25G
TCIAB1948-5G
3-Bromo-5-chloro-2-hydroxybenzaldehyde ≥96.0% (by GC)
3-Bromo-5-chloro-2-hydroxybenzaldehyde
Formula:
C₇H₄BrClO₂ Peso molecolare: 235,46 g/mol |
MDL Number:
MFCD00051690 CAS Number: 19652-32-5 |
Risultati del test di specificazione
Appearance | White to Light yellow to Green powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 85.0 to 89.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...