2-Ethylhexyl methacrylate ≥99.0% (by GC) stabilizzato
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Methacrylic acid 2-ethylhexyl ester
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
M0591-25ML
M0591-500ML
TCIAM0591-25MLEA
18
EUR
TCIAM0591-25ML
TCIAM0591-500ML
2-Ethylhexyl methacrylate ≥99.0% (by GC) stabilizzato
2-Ethylhexyl methacrylate
Formula:
C₁₂H₂₂O₂ Peso molecolare: 198,31 g/mol Punto di ebollizione: 88°C Flash Pt: 100°C |
MDL Number:
MFCD00009494 CAS Number: 688-84-6 EINECS: 211-708-6 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...