2,3-Dimethylcyclohexanol (mixture of isomers) ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
2,3-Dimethylcyclohexanol
D1660-25ML
D1660-5ML
TCIAD1660-25MLEA
173
EUR
TCIAD1660-25ML
TCIAD1660-5ML
2,3-Dimethylcyclohexanol (mixture of isomers) ≥98.0% (by GC)
2,3-Dimethylcyclohexanol (mixture of isomers)
Formula:
C₈H₁₆O Peso molecolare: 128,21 g/mol Punto di ebollizione: 181°C Densità: 0,93 g/cm³ (20 °C) Flash Pt: 66°C |
MDL Number:
MFCD00001444 CAS Number: 1502-24-5 EINECS: 216-121-9 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % (total of isomers) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...