3-Ammino-9-etil carbazolo (AEC) ≥90.0% (by HPLC, titration analysis) for biochemical research
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
9-Etilcarbazol-3-ilammina, AEC
A2167-1G
A2167-5G
TCIAA2167-1GEA
20
EUR
TCIAA2167-1G
TCIAA2167-5G
3-Ammino-9-etil carbazolo (AEC) ≥90.0% (by HPLC, titration analysis) for biochemical research
3-Ammino-9-etil carbazolo (AEC)
Formula:
C₁₄H₁₄N₂ Peso molecolare: 210,28 g/mol Punto di fusione: 113…114 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00004964 CAS Number: 132-32-1 EINECS: 205-057-7 UN: 2811 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (HPLC) | min. 90.0 area% |
Purity (Nonaqueous Titration) | min. 90.0 % |
Peroxidase detection test | to pass test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...