Stampa…

Furan-2-boronic acid (contains varying amounts of Anhydride)

Fornitore: TCI
Attenzione

Synonyms: Furan-2-ylboronic acid

F0394-1G F0394-5G
TCIAF0394-1GEA 24 EUR
TCIAF0394-1G TCIAF0394-5G
Furan-2-boronic acid (contains varying amounts of Anhydride)
Furan-2-boronic acid
Formula: C₄H₅BO₃
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00799544
CAS Number: 13331-23-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d398a0ffa9dd3505343d814e60157c9f23334a9&vid=6d398a0ffa9dd3505343d814e60157c9f23334a9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (Neutralization titration) 97.0 to 119.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR