2-Fluoro-3-pyridineboronic acid (contains varying amounts of Anhydride)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Fluoro-3-pyridinylboronic acid, (2-Fluoropyridin-3-yl)boronic acid, 2-Fluoropyridine-3-boronic acid
F0739-25G
F0739-5G
TCIAF0739-25GEA
336
EUR
TCIAF0739-25G
TCIAF0739-5G
2-Fluoro-3-pyridineboronic acid (contains varying amounts of Anhydride)
2-Fluoro-3-pyridineboronic acid
Formula:
C₅H₅BFNO₂ Peso molecolare: 140,91 g/mol Punto di fusione: 172°C |
MDL Number:
MFCD03411557 CAS Number: 174669-73-9 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 98.0 to 115.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...