2,2'-Thiodiacetic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Synonyms:
Thiodiglycolic acid, Thiodiacetic Acid
T1248-25G
T1248-500G
TCIAT1248-25GEA
23
EUR
TCIAT1248-25G
TCIAT1248-500G
2,2'-Thiodiacetic acid ≥98.0% (by GC, titration analysis)
2,2'-Thiodiacetic acid
Formula:
C₄H₆O₄S Peso molecolare: 150,16 g/mol Punto di ebollizione: 168 °C (14 -rr) Punto di fusione: 128…131 °C Densità: 1,661 g/cm³ (20 °C) |
MDL Number:
MFCD00004359 CAS Number: 123-93-3 EINECS: 204-663-9 UN: 3261 ADR: 8,III Merck Index: 14,09331 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 128.0 to 132.0 deg-C |
Solubility in Water | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...