Stampa…

5-Metilfenazoniometilsolfato ≥98.0% (by HPLC, total nitrogen) for biochemical research

Fornitore: TCI EUROPE
Attenzione

Synonyms: PMS, 5-Methylphenazin-5-ium methyl sulphate, Phenazine methosulphate, Phenazine methylsulphate

P1872-1G P1872-5G
TCIAP1872-1GEA 27 EUR
TCIAP1872-1G TCIAP1872-5G
5-Metilfenazoniometilsolfato ≥98.0% (by HPLC, total nitrogen) for biochemical research
5-Methylphenazinium methyl sulphate
Formula: C₁₄H₁₄N₂O₄S
Peso molecolare: 306,34 g/mol
Punto di fusione: 160°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00011923
CAS Number: 299-11-6
EINECS: 206-072-1
Merck Index: 14,06109
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22dfd45999ff362c6592f1cc0a0720851722b483&vid=5bb413d32272a7942ba921d8fa10d4075af963df&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Enzyme detection test to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR