1-Ethyl-3-methylimidazoliumtetrafluoroborate ≥97.0% (by total nitrogen basis) for molten salt
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
1-Ethyl-3-methyl-1H-imidazol-3-ium tetrafluoroborate
E0496-25G
E0496-5G
TCIAE0496-25GEA
148
EUR
TCIAE0496-25G
TCIAE0496-5G
1-Ethyl-3-methylimidazoliumtetrafluoroborate ≥97.0% (by total nitrogen basis) for molten salt
1-Ethyl-3-methylimidazoliumtetrafluoroborate
Formula:
C₆H₁₁BF₄N₂ Peso molecolare: 197,97 g/mol Punto di ebollizione: ? 200 °C (1013 hPa) Punto di fusione: 15°C Densità: 1,34 g/cm³ (20 °C) Flash Pt: ≥ 110 °C |
MDL Number:
MFCD00216668 CAS Number: 143314-16-3 UN: 1760 ADR: 8,III |
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (HPLC) | min. 95.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...