Stampa…

4-Amminoantipirina ≥98.0% (by titrimetric analysis) for biochemical research

Fornitore: TCI
Attenzione

Synonyms: 4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Ampyrone, 4-Aminophenazone

A2254-1G A2254-5G
TCIAA2254-1GEA 16 EUR
TCIAA2254-1G TCIAA2254-5G
4-Amminoantipirina ≥98.0% (by titrimetric analysis) for biochemical research
4-Amminoantipirina
Formula: C₁₁H₁₃N₃O
Peso molecolare: 203.24 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00003145
CAS Number: 83-07-8
EINECS: 201-452-3
Merck Index: 14,00591
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c8d644bd64bc916afd15d4ee016e47da6718bef&vid=103d4393579d3958fad24c16e6b6837715df1aa8&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 107.0 to 111.0 deg-C
Hydrogendioxide detection test (Peroxidase method) to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR