Acido 2-[Bis(2-idrossietil)amino]etansolfonico ≥98.0% (by titrimetric analysis) good's buffer component for biological research
Fornitore: TCI
|
Attenzione
|
Synonyms:
BES, 2-(Bis(2-hydroxyethyl)amino)ethanesulphonic acid, N,N-Bis-(2-hydroxyethyl)-2-ethanesulphonic acid
Formula:
C₆H₁₅NO₅S Peso molecolare: 213.25 g/mol Punto di ebollizione: 405 Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00007533 CAS Number: 10191-18-1 EINECS: 233-465-5 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Absorbance | max. 0.15 (1mol/L、H2O、260nm) |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...