Stampa…

Pentacene, purified by sublimation

Fornitore: TCI EUROPE

Synonyms: 2,3,6,7-Dibenzanthracene

P0030-1G P0030-100MG
TCIAP0030-1GEA 244 EUR
TCIAP0030-1G TCIAP0030-100MG
Pentacene, purified by sublimation
Pentacene
Formula: C₂₂H₁₄
Peso molecolare: 278,35 g/mol
Punto di fusione: 300 °C
Densità: 1,3 g/cm³ (20 °C)
MDL Number: MFCD00003710
CAS Number: 135-48-8
EINECS: 205-193-7
Merck Index: 14,07107
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1fcca75eca7c6ca50aaf4678d80c520f67766afd&vid=ec5ca7a202e7d990135e840d8470a6c2d2e00190&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Gray to Dark purple to Black powder to crystal
TOF-MS to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR