4-((Trifluoromethyl)thio)benzoic acid ≥97.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
T2390-25G
T2390-5G
TCIAT2390-25GEA
189
EUR
TCIAT2390-25G
TCIAT2390-5G
4-((Trifluoromethyl)thio)benzoic acid ≥97.0% (by GC, titration analysis)
4-((Trifluoromethyl)thio)benzoic acid
Formula:
C₈H₅F₃O₂S Peso molecolare: 222.19 g/mol |
MDL Number:
MFCD00040906 CAS Number: 330-17-6 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 159.0 to 163.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...