N,N-Dimethylformamide dibutyl acetal ≥98.0% (by GC, titration analysis) for esterification
Fornitore: TCI
|
Attenzione
|
D1302-25ML
D1302-5ML
TCIAD1302-25MLEA
108
EUR
TCIAD1302-25ML
TCIAD1302-5ML
N,N-Dimethylformamide dibutyl acetal ≥98.0% (by GC, titration analysis) for esterification
N,N-Dimethylformamide dibutyl acetal
Formula:
C₁₁H₂₅NO₂ Punto di ebollizione: 98°C Densità: 0,852 g/cm³ (20 °C) Flash Pt: 53°C |
MDL Number:
MFCD00015249 CAS Number: 18503-90-7 EINECS: 242-387-0 UN: 1993 ADR: 3,III |
|
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Effect test of GC derivalization | Effective as derivatization agent Myristic Acid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...