4-Hydroxyphenylboronic acid (contains varying amounts of Anhydride)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(4-Hydroxyphenyl)metaboric acid, Dihydroxy(4-hydroxyphenyl)borane, 4-Hydroxybenzeneboronic acid
H1228-1G
H1228-5G
TCIAH1228-1GEA
33
EUR
TCIAH1228-1G
TCIAH1228-5G
4-Hydroxyphenylboronic acid (contains varying amounts of Anhydride)
4-Hydroxyphenylboronic acid
Formula:
C₆H₇BO₃ Peso molecolare: 137,93 g/mol Punto di fusione: 232°C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD01074628 CAS Number: 71597-85-8 |
Risultati del test di specificazione
Appearance | White to Gray to Brown powder to crystal |
Purity (Neutralization titration) | 97.0 to 115.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...