Stampa…

4-Fluorophenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (4-Fluorophenyl)metaboric acid, 4-Fluorobenzeneboronic acid, Dihydroxy(4-fluorophenyl)borane

F0361-25G F0361-5G
TCIAF0361-25GEA 78 EUR
TCIAF0361-25G TCIAF0361-5G
4-Fluorophenylboronic acid (contains varying amounts of Anhydride)
4-Fluorophenylboronic acid
Formula: C₆H₆BFO₂
Peso molecolare: 139,92 g/mol
Punto di fusione: 263…265 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00039136
CAS Number: 1765-93-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00b4d3bdffe3302c08525b8da8f64ecf84a7ad1d&vid=00b4d3bdffe3302c08525b8da8f64ecf84a7ad1d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light red powder to crystal
Purity (Neutralization titration) 97.0 to 116.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR